Learn More
Grainger Thread Mill Carbide
Catalog No. 502764959
Encompass_Preferred
The Micro 100 16R742 is a carbide thread mill with a 1/2" shank diameter, 1-1/2" overall length, and 1/4"-28 internal thread size. It has a TiAlN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its variable helix flutes, it provides efficient chip evacuation while threading, allowing for optimal performance. The Micro 100 thread mill is made in the USA and well-suited for production threading applications.
Catalog No. 50-276-4959 Supplier Grainger Supplier No. 16R742
The Micro 100 16R742 is a carbide thread mill with a 1/2" shank diameter, 1-1/2" overall length, and 1/4"-28 internal thread size. It has a TiAlN coating for increased tool life and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. With its variable helix flutes, it provides efficient chip evacuation while threading, allowing for optimal performance. The Micro 100 thread mill is made in the USA and well-suited for production threading applications.