Learn More
Grainger Thread Mill Carbide
Catalog No. 502765841
Encompass_Preferred
The Thread Mill Carbide 16R762 is a solid carbide thread milling cutter manufactured by EFC. It has a 16 mm diameter, can machine a variety of thread sizes including M6, M8, M10, and M12, and features an AlTiN coating for increased tool life and performance. The cutter has 6 flutes and can machine threads in steel, stainless steel, cast iron, aluminum, and other materials. It has a right-hand cut direction. The 16R762 thread mill is well-suited for difficult to machine materials and for blind holes and cavities. It provides an efficient method of thread milling.
Catalog No. 50-276-5841 Supplier Grainger Supplier No. 16R762
The Thread Mill Carbide 16R762 is a solid carbide thread milling cutter manufactured by EFC. It has a 16 mm diameter, can machine a variety of thread sizes including M6, M8, M10, and M12, and features an AlTiN coating for increased tool life and performance. The cutter has 6 flutes and can machine threads in steel, stainless steel, cast iron, aluminum, and other materials. It has a right-hand cut direction. The 16R762 thread mill is well-suited for difficult to machine materials and for blind holes and cavities. It provides an efficient method of thread milling.