Learn More
Grainger Thread Mill Carbide
Catalog No. 502765842
Encompass_Preferred
The Micro 100 16R757 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads from 10-56 to 1/4-20. The overall length is 2-1/2" and it has a 1/4" shank. This thread mill is made in the USA and is designed for high performance threading in steels, stainless steels, cast iron, aluminum and other materials. It features Micro 100's proprietary TiAlN coating for increased tool life and improved performance. The 16R757 thread mill is well-suited for production threading applications.
Catalog No. 50-276-5842 Supplier Grainger Supplier No. 16R757
The Micro 100 16R757 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads from 10-56 to 1/4-20. The overall length is 2-1/2" and it has a 1/4" shank. This thread mill is made in the USA and is designed for high performance threading in steels, stainless steels, cast iron, aluminum and other materials. It features Micro 100's proprietary TiAlN coating for increased tool life and improved performance. The 16R757 thread mill is well-suited for production threading applications.