Learn More
Grainger Thread Mill Carbide
Catalog No. 502764960
Encompass_Preferred
The Micro 100 16R732 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree internal threads. The overall length is 2-1/2" and it has a 1/2" shank. The thread mill has a reach of 1-1/4" and can cut thread depths up to 1X diameter. It is capable of milling a wide range of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The 16R732 thread mill is manufactured in the USA by Micro 100 and is well-suited for production threading applications.
Catalog No. 50-276-4960 Supplier Grainger Supplier No. 16R732
The Micro 100 16R732 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree internal threads. The overall length is 2-1/2" and it has a 1/2" shank. The thread mill has a reach of 1-1/4" and can cut thread depths up to 1X diameter. It is capable of milling a wide range of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics and more. The 16R732 thread mill is manufactured in the USA by Micro 100 and is well-suited for production threading applications.