Learn More
Grainger Thread Mill Carbide
Catalog No. 502766155
Encompass_Preferred
The Micro 100 16R739 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 2-1/2" and it has a 1/2" shank. This thread mill is designed for high feed milling in steels, tool steels, stainless steels and cast iron. It can be run at speeds up to 2000 SFM.
Catalog No. 50-276-6155 Supplier Grainger Supplier No. 16R739
The Micro 100 16R739 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads. The overall length is 2-1/2" and it has a 1/2" shank. This thread mill is designed for high feed milling in steels, tool steels, stainless steels and cast iron. It can be run at speeds up to 2000 SFM.