Learn More
Grainger Thread Mill Carbide
Catalog No. 502766158
Encompass_Preferred
The Micro 100 16R731 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree internal threads. The overall length is 2-1/2" and it has a 1/2" shank. This thread mill is designed for use on steel, stainless steel, cast iron, aluminum and other non-ferrous metals. It can be run at speeds up to 2000 SFM.
Catalog No. 50-276-6158 Supplier Grainger Supplier No. 16R731
The Micro 100 16R731 is a 1/4" diameter, 4 flute, carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree internal threads. The overall length is 2-1/2" and it has a 1/2" shank. This thread mill is designed for use on steel, stainless steel, cast iron, aluminum and other non-ferrous metals. It can be run at speeds up to 2000 SFM.