Learn More
Grainger Thread Mill Carbide
Catalog No. 502766164
Encompass_Preferred
The Grainger 16R727 is a Westward 3/8-inch drive deep impact socket set. This 12-piece socket set includes sizes ranging from 10mm to 19mm, providing a versatile range of metric sockets for impact wrench use. The sockets are made from chrome vanadium steel for durability and feature a black phosphate finish to resist corrosion. With deep broached hexagons that grip fasteners tightly and ABS plastic storage cases to keep the sockets organized, this Westward impact socket set is designed for demanding automotive repair applications.
Catalog No. 50-276-6164 Supplier Grainger Supplier No. 16R727
The Grainger 16R727 is a Westward 3/8-inch drive deep impact socket set. This 12-piece socket set includes sizes ranging from 10mm to 19mm, providing a versatile range of metric sockets for impact wrench use. The sockets are made from chrome vanadium steel for durability and feature a black phosphate finish to resist corrosion. With deep broached hexagons that grip fasteners tightly and ABS plastic storage cases to keep the sockets organized, this Westward impact socket set is designed for demanding automotive repair applications.