Learn More
Grainger Thread Mill Carbide
Catalog No. 502767466
Encompass_Preferred
The Grainger 16R652 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench. It has a working torque range up to 250 ft-lbs. The overall length of the adapter is 2-1/2 inches.
Catalog No. 50-276-7466 Supplier Grainger Supplier No. 16R652
The Grainger 16R652 is a Westward 3/8-inch drive impact socket adapter that converts a 3/8-inch drive to a 1/2-inch drive. It is made from chrome vanadium steel for durability and has a black phosphate finish to resist corrosion. This impact socket adapter allows you to use 3/8-inch drive tools with a 1/2-inch drive impact wrench. It has a working torque range up to 250 ft-lbs. The overall length of the adapter is 2-1/2 inches.