Learn More
Grainger Thread Mill Carbide
Catalog No. 502768327
Encompass_Preferred
The Micro 100 16R756 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a right hand spiral flute. It has a TiAlN coating and can cut 60-62 HRC materials. The thread mill has a 1/4"-28 UNF shank and can cut thread sizes from #10-24 through 1/4"-28. It has a neck relief below the thread for improved chip clearance. The Micro 100 16R756 thread mill is made in the USA.
Catalog No. 50-276-8327 Supplier Grainger Supplier No. 16R756
The Micro 100 16R756 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a right hand spiral flute. It has a TiAlN coating and can cut 60-62 HRC materials. The thread mill has a 1/4"-28 UNF shank and can cut thread sizes from #10-24 through 1/4"-28. It has a neck relief below the thread for improved chip clearance. The Micro 100 16R756 thread mill is made in the USA.