Learn More
Grainger Thread Mill Carbide
Catalog No. 502768392
Encompass_Preferred
The Micro 100 16R750 is a 1/2" diameter, 4 flute, right hand thread mill made of solid carbide with TiAlN coating. It has a shank diameter of 3/8", an overall length of 2-1/2", and can cut unified thread sizes from 4-48 to 1/2-13. The helix angle is 30 degrees. This thread mill is designed for high feed milling applications in steels, stainless steels, cast iron, aluminum, and other materials. It features a variable helix flute design to reduce harmonics and provide a smooth cut. The TiAlN coating provides heat and wear resistance. Made in USA by Micro 100 Tool Corp.
Catalog No. 50-276-8392 Supplier Grainger Supplier No. 16R750
The Micro 100 16R750 is a 1/2" diameter, 4 flute, right hand thread mill made of solid carbide with TiAlN coating. It has a shank diameter of 3/8", an overall length of 2-1/2", and can cut unified thread sizes from 4-48 to 1/2-13. The helix angle is 30 degrees. This thread mill is designed for high feed milling applications in steels, stainless steels, cast iron, aluminum, and other materials. It features a variable helix flute design to reduce harmonics and provide a smooth cut. The TiAlN coating provides heat and wear resistance. Made in USA by Micro 100 Tool Corp.