Learn More
Grainger Thread Mill Carbide
Catalog No. 502765341
Encompass_Preferred
The Micro 100 16R763 is a 1/4" diameter, 3/4" overall length, 4 flute thread mill with a TiAlN coating. It has a right hand spiral flute direction and can cut 60 degree threads. The thread mill has a shank diameter of 3/8" and a reach of 5/8". It is made of solid carbide and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper alloys, plastics and composites. The 16R763 thread mill is designed for high performance threading applications.
Catalog No. 50-276-5341 Supplier Grainger Supplier No. 16R763
The Micro 100 16R763 is a 1/4" diameter, 3/4" overall length, 4 flute thread mill with a TiAlN coating. It has a right hand spiral flute direction and can cut 60 degree threads. The thread mill has a shank diameter of 3/8" and a reach of 5/8". It is made of solid carbide and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper alloys, plastics and composites. The 16R763 thread mill is designed for high performance threading applications.