Learn More
Grainger Thread Mill Carbide
Catalog No. 502765343
Encompass_Preferred
The Micro 100 16R774 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads from 10-56 to 1/4-20. The overall length is 3.75" and it has a 1/4" shank. This thread mill is made in the USA out of micrograin carbide with titanium aluminum nitride coating for wear resistance when cutting steel, stainless steel, cast iron, aluminum and other materials. It can be run at speeds up to 2000 SFM.
Catalog No. 50-276-5343 Supplier Grainger Supplier No. 16R774
The Micro 100 16R774 is a 1/4" diameter, 3/4" long, 4 flute carbide thread mill with a TiAlN coating. It has a right hand spiral flute and can cut 60 degree threads from 10-56 to 1/4-20. The overall length is 3.75" and it has a 1/4" shank. This thread mill is made in the USA out of micrograin carbide with titanium aluminum nitride coating for wear resistance when cutting steel, stainless steel, cast iron, aluminum and other materials. It can be run at speeds up to 2000 SFM.