Learn More
Chemglass Life Sciences Chem-Thread, Size 7, 11mm Tube OD, Accepts 4-7mm OD Tubing, #107 O-ring
SDP
Catalog No. 50873751
Encompass_Preferred
Chem-Thread, Size 7, 11mm Tube OD, Accepts 4-7mm OD Tubing, #107 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.
Catalog No. 50-873-751 Supplier Chemglass Life Sciences Supplier No. CG35001
Chem-Thread, Size 7, 11mm Tube OD, Accepts 4-7mm OD Tubing, #107 O-ring. Consists of an externally threaded glass tube, Viton o-ring and Celcon compression cap. Tube O.D. listed is the shank O.D. of the connector.