Learn More
MSC Balax Thread Plug Gage, #1-64 Thread, 2B and 3B Class, Go, 1 ea.
Catalog No. 5000516877
Encompass
This single-end go thread plug gage is designed for precise inspection of internal threads, ensuring conformity to #1-64 thread specifications. Constructed from high speed tool steel, this tool is engineered for durability and consistent performance in industrial measurement applications.
  • Features a #1-64 thread size compatible with 2B and 3B classes
  • Constructed from high speed tool steel with a 63-65 Rockwell hardness rating
  • Equipped with a #000 taperlock handle
  • Provides a 0.25 in. thread length and 0.9375 in. (15/16 in.) overall length
  • Finished with a bright coating
Catalog No. 50-005-16877 Supplier MSC Supplier No. 05351648
This single-end go thread plug gage is designed for precise inspection of internal threads, ensuring conformity to #1-64 thread specifications. Constructed from high speed tool steel, this tool is engineered for durability and consistent performance in industrial measurement applications.
  • Features a #1-64 thread size compatible with 2B and 3B classes
  • Constructed from high speed tool steel with a 63-65 Rockwell hardness rating
  • Equipped with a #000 taperlock handle
  • Provides a 0.25 in. thread length and 0.9375 in. (15/16 in.) overall length
  • Finished with a bright coating