Learn More
MSC Balax Thread Plug Gage, #0-80, 3B Class, Double End, 1 ea
Catalog No. 5000512705
Encompass
This double end thread plug gage is designed for precise inspection of #0-80 internal threads with a 3B class of fit. Constructed for durability and accuracy in industrial applications, this gage features go and no-go ends to ensure threads meet stringent quality standards.
  • Constructed from high speed tool steel for enhanced wear resistance
  • Features 63-65 Rockwell hardness for extended service life
  • Designed for 3B class of fit UNF thread classification
  • Equipped with a #000 taperlock handle size
  • Bright finish for high visibility and corrosion resistance
Catalog No. 50-005-12705 Supplier MSC Supplier No. 05360839
This double end thread plug gage is designed for precise inspection of #0-80 internal threads with a 3B class of fit. Constructed for durability and accuracy in industrial applications, this gage features go and no-go ends to ensure threads meet stringent quality standards.
  • Constructed from high speed tool steel for enhanced wear resistance
  • Features 63-65 Rockwell hardness for extended service life
  • Designed for 3B class of fit UNF thread classification
  • Equipped with a #000 taperlock handle size
  • Bright finish for high visibility and corrosion resistance