Learn More
MSC GF Gage Thread Plug Gage, 2-1/2-12, 2B, 1 ea
Catalog No. 5000548002
Encompass
This thread plug gage is designed for checking the accuracy of 2-1/2-12 2B internal threads. It features a GO/NO GO configuration, allowing for efficient verification of pitch diameter and thread form compliance within specified tolerances.
  • Designed for 2-1/2-12 2B thread specifications
  • Includes both GO and NO GO members for comprehensive inspection
  • Precision construction ensures reliable thread verification
Catalog No. 50-005-48002 Supplier MSC Supplier No. 53189551
This thread plug gage is designed for checking the accuracy of 2-1/2-12 2B internal threads. It features a GO/NO GO configuration, allowing for efficient verification of pitch diameter and thread form compliance within specified tolerances.
  • Designed for 2-1/2-12 2B thread specifications
  • Includes both GO and NO GO members for comprehensive inspection
  • Precision construction ensures reliable thread verification