Learn More
MSC Balax Thread Plug Gage, #3-48, 2B and 3B Class, Go, 1 ea
Catalog No. 5000511155
Encompass
This thread plug gage is designed for the inspection of internal threads, providing a reliable way to verify that parts meet specific tolerance requirements. It features a high speed tool steel construction with a bright finish for durability and precision in metalworking applications.
  • Designed with a #3-48 thread size
  • Compatible with 2B and 3B class of fit
  • Single end GO member configuration
  • Constructed from high speed tool steel
  • Hardness rated at 63 to 65 Rockwell
  • Overall length of 15/16 in. with a thread length of 1/4 in.
  • Equipped with a #000 taperlock handle
Catalog No. 50-005-11155 Supplier MSC Supplier No. 05353487
This thread plug gage is designed for the inspection of internal threads, providing a reliable way to verify that parts meet specific tolerance requirements. It features a high speed tool steel construction with a bright finish for durability and precision in metalworking applications.
  • Designed with a #3-48 thread size
  • Compatible with 2B and 3B class of fit
  • Single end GO member configuration
  • Constructed from high speed tool steel
  • Hardness rated at 63 to 65 Rockwell
  • Overall length of 15/16 in. with a thread length of 1/4 in.
  • Equipped with a #000 taperlock handle