Learn More
MSC Balax Thread Plug Gage, #8-36, 2B and 3B Class, Go, 1 ea.
Catalog No. 5000510973
Encompass
This single-end thread plug gage is designed for precise mechanical thread inspection to verify that workpieces conform to specified tolerances. Constructed from hardened high-speed tool steel, this instrument provides durability and consistent measurement accuracy for demanding manufacturing environments.
  • Designed for #8-36 thread sizes
  • Compatible with 2B and 3B tolerance classes
  • Hardened to 63-65 Rockwell C
  • Features a #0 taperlock handle size
  • Includes 13/32 in. thread length and 1-5/8 in. overall length
  • Suitable for production machining and quality control applications
Catalog No. 50-005-10973 Supplier MSC Supplier No. 05357363
This single-end thread plug gage is designed for precise mechanical thread inspection to verify that workpieces conform to specified tolerances. Constructed from hardened high-speed tool steel, this instrument provides durability and consistent measurement accuracy for demanding manufacturing environments.
  • Designed for #8-36 thread sizes
  • Compatible with 2B and 3B tolerance classes
  • Hardened to 63-65 Rockwell C
  • Features a #0 taperlock handle size
  • Includes 13/32 in. thread length and 1-5/8 in. overall length
  • Suitable for production machining and quality control applications