Learn More
MSC GF Gage Threaded Ring Gage, 7/8-9 UNC, Class 3A, Go and No Go, 1 ea
Catalog No. 5000511459
Encompass
This threaded ring gage set is designed for precise inspection of external thread dimensions, featuring both Go and No Go components to ensure compliance with quality standards. It is engineered for durability and accuracy in rigorous metalworking and inspection environments.
  • Designed for 7/8-9 UNC threads
  • Class 3A classification
  • Includes both Go and No Go gages
  • Double ring configuration
  • Manufactured in the United States
Catalog No. 50-005-11459 Supplier MSC Supplier No. 55326938
This threaded ring gage set is designed for precise inspection of external thread dimensions, featuring both Go and No Go components to ensure compliance with quality standards. It is engineered for durability and accuracy in rigorous metalworking and inspection environments.
  • Designed for 7/8-9 UNC threads
  • Class 3A classification
  • Includes both Go and No Go gages
  • Double ring configuration
  • Manufactured in the United States