Learn More
MSC GF Gage Threaded Ring Gage, 1-3/8-6 UNC, Class 2A, 1 ea.
Catalog No. 5000511403
Encompass
This threaded ring gage is designed for precision inspection of external threads, featuring both Go and No Go components to verify thread size and tolerance. It is suited for applications requiring high-accuracy thread measurement in manufacturing and quality control environments.
  • Includes a 2-piece set for Go and No Go verification
  • Constructed to 2A classification standards
  • Designed for 1-3/8-6 UNC thread specifications
  • Double ring configuration
Catalog No. 50-005-11403 Supplier MSC Supplier No. 55325997
This threaded ring gage is designed for precision inspection of external threads, featuring both Go and No Go components to verify thread size and tolerance. It is suited for applications requiring high-accuracy thread measurement in manufacturing and quality control environments.
  • Includes a 2-piece set for Go and No Go verification
  • Constructed to 2A classification standards
  • Designed for 1-3/8-6 UNC thread specifications
  • Double ring configuration