Learn More
MSC GF Gage Threaded Ring Gage, 3/8-32 UNEF, Class 2A, 1 ea
Catalog No. 5000512216
Encompass
This threaded ring gage set is designed for the precise measurement and inspection of external threads. The set includes both Go and No Go gages to ensure threads meet the specified tolerances for industrial applications.
  • Thread size is 3/8-32
  • Thread type is UNEF
  • Classification is 2A
  • Double ring configuration
  • Includes Go and No Go gages
Catalog No. 50-005-12216 Supplier MSC Supplier No. 55325450
This threaded ring gage set is designed for the precise measurement and inspection of external threads. The set includes both Go and No Go gages to ensure threads meet the specified tolerances for industrial applications.
  • Thread size is 3/8-32
  • Thread type is UNEF
  • Classification is 2A
  • Double ring configuration
  • Includes Go and No Go gages