Learn More
MSC GF Gage Thread Ring Gage, #10-32 UNF, Class 3A, No Go, 1 ea.
Catalog No. 5000511402
Encompass
This threaded ring gage is designed for precise inspection of #10-32 UNF threads. This No Go gage ensures that manufactured parts meet the strict requirements of Class 3A tolerance standards.
  • Designed for No Go inspection of threaded components
  • Threads to #10-32 UNF specification
  • Class 3A classification for high-precision measurement
  • Single ring configuration
Catalog No. 50-005-11402 Supplier MSC Supplier No. 55324073
This threaded ring gage is designed for precise inspection of #10-32 UNF threads. This No Go gage ensures that manufactured parts meet the strict requirements of Class 3A tolerance standards.
  • Designed for No Go inspection of threaded components
  • Threads to #10-32 UNF specification
  • Class 3A classification for high-precision measurement
  • Single ring configuration