Learn More
MSC GF Gage Threaded Ring Gage, 1/4-28, UNF, 2A, Tool Steel, 1 ea.
Catalog No. 5000514540
Encompass
This threaded ring gage is designed for precision inspection of external threads. Manufactured from durable tool steel, this single-ring go gage provides reliable verification for 1/4-28 UNF threads.
  • Constructed from high-quality tool steel for durability
  • Designed for 1/4-28 UNF thread specifications
  • Class 2A and 6G rating for precise inspection
  • Go configuration for efficient thread verification
Catalog No. 50-005-14540 Supplier MSC Supplier No. 55320675
This threaded ring gage is designed for precision inspection of external threads. Manufactured from durable tool steel, this single-ring go gage provides reliable verification for 1/4-28 UNF threads.
  • Constructed from high-quality tool steel for durability
  • Designed for 1/4-28 UNF thread specifications
  • Class 2A and 6G rating for precise inspection
  • Go configuration for efficient thread verification