Learn More
MSC GF Gage Thread Ring Gage, 7/16-28 Thread, Class 3A, No Go, 1 ea
Catalog No. 5000514541
Encompass
This threaded ring gage is designed for precision inspection of external threads, specifically providing a reliable method for checking the No Go status of parts. It is engineered to meet 3A class specifications, ensuring accurate and consistent measurement performance in industrial metalworking applications.
  • Designed for 7/16-28 thread size
  • Class 3A classification for high-precision inspection
  • No Go configuration for effective thread validation
  • Manufactured for consistent dimensional accuracy
Catalog No. 50-005-14541 Supplier MSC Supplier No. 55324354
This threaded ring gage is designed for precision inspection of external threads, specifically providing a reliable method for checking the No Go status of parts. It is engineered to meet 3A class specifications, ensuring accurate and consistent measurement performance in industrial metalworking applications.
  • Designed for 7/16-28 thread size
  • Class 3A classification for high-precision inspection
  • No Go configuration for effective thread validation
  • Manufactured for consistent dimensional accuracy