Learn More
MSC GF Gage Threaded Ring Gage, #5-40 Thread, Class 3A, Go, 1 ea
Catalog No. 5000513935
Encompass
This threaded ring gage is designed for precision inspection of #5-40 thread size fasteners to ensure they meet 3A class specifications.
  • Go gage configuration for reliable thread verification
  • Single ring design for efficient use
  • Class 3A classification for high-precision thread tolerance requirements
Catalog No. 50-005-13935 Supplier MSC Supplier No. 55321616
This threaded ring gage is designed for precision inspection of #5-40 thread size fasteners to ensure they meet 3A class specifications.
  • Go gage configuration for reliable thread verification
  • Single ring design for efficient use
  • Class 3A classification for high-precision thread tolerance requirements