Learn More
MSC GF Gage Threaded Ring Gage, #12-24, Class 3A, No Go, 1 ea
Catalog No. 5000513937
Encompass
This threaded ring gage is designed for precision inspection of external threads to ensure they meet specified tolerance requirements. It serves as a reliable tool for verifying thread dimensions during manufacturing and quality control processes.
  • Thread size: #12-24
  • Classification: 3A
  • Type: No go
  • Configuration: Single ring
Catalog No. 50-005-13937 Supplier MSC Supplier No. 55324099
This threaded ring gage is designed for precision inspection of external threads to ensure they meet specified tolerance requirements. It serves as a reliable tool for verifying thread dimensions during manufacturing and quality control processes.
  • Thread size: #12-24
  • Classification: 3A
  • Type: No go
  • Configuration: Single ring