Learn More
MSC GF Gage Thread Ring Gage, #2-56, UNC, 2A, No Go, 1 ea
Catalog No. 5000513938
Encompass
This threaded ring gage is designed for precise inspection of external threads, ensuring they meet specified tolerances. It is a no go gage used to verify that the maximum pitch diameter of the thread is not exceeded.
  • Thread size of #2-56
  • UNC thread type
  • Class 2A classification
  • No go gage type
  • Single ring configuration
Catalog No. 50-005-13938 Supplier MSC Supplier No. 55322655
This threaded ring gage is designed for precise inspection of external threads, ensuring they meet specified tolerances. It is a no go gage used to verify that the maximum pitch diameter of the thread is not exceeded.
  • Thread size of #2-56
  • UNC thread type
  • Class 2A classification
  • No go gage type
  • Single ring configuration