Learn More
MSC Gf Gage Threaded Ring Gage, #8-36 Thread, Class 3A, Go, 1 ea
Catalog No. 5000512212
Encompass
This threaded ring gage is designed for precision inspection of external threads. It is a single ring gage specifically manufactured to verify the accuracy of #8-36 threads according to Class 3A tolerance standards.
  • Designed as a Go gage for precise thread inspection
  • Compatible with #8-36 thread sizes
  • Maintains Class 3A tolerance standards
  • Single ring gage construction
Catalog No. 50-005-12212 Supplier MSC Supplier No. 55321715
This threaded ring gage is designed for precision inspection of external threads. It is a single ring gage specifically manufactured to verify the accuracy of #8-36 threads according to Class 3A tolerance standards.
  • Designed as a Go gage for precise thread inspection
  • Compatible with #8-36 thread sizes
  • Maintains Class 3A tolerance standards
  • Single ring gage construction