Learn More
MSC GF Gage Threaded Ring Gage, #5-44 Thread, Class 3A, Go, 1 ea
Catalog No. 5000512018
Encompass
This threaded ring gage is designed for precision inspection of external threads, ensuring they meet specified size and fit requirements. It is a single-ring configuration engineered for accurate measurement in metalworking and machining applications.
  • Class 3A thread classification
  • Number 5-44 thread size
  • Go type gage
Catalog No. 50-005-12018 Supplier MSC Supplier No. 55321632
This threaded ring gage is designed for precision inspection of external threads, ensuring they meet specified size and fit requirements. It is a single-ring configuration engineered for accurate measurement in metalworking and machining applications.
  • Class 3A thread classification
  • Number 5-44 thread size
  • Go type gage