Learn More
Perkin Elmer US LLC Thread Sealant Tape
Catalog No. 502113079
Encompass_Preferred
This multipupose industrial tape is compatible with TMA and TGA instruments and accessories. It ships standard with a variety of the installation and maintenance kits for these instruments.
Catalog No. 50-211-3079 Supplier Perkin Elmer US LLC Supplier No. 09908134
This multipupose industrial tape is compatible with TMA and TGA instruments and accessories. It ships standard with a variety of the installation and maintenance kits for these instruments.