Learn More
Chemglass Life Sciences Thread Sealant, with PTFE, 2oz Tube
SDP
Catalog No. 501311161
Encompass_Preferred
Thread Sealant, with PTFE, 2oz Tube. Smooth, nontoxic, paste forms a leak tight seal on pipe threads. Ideal for use with Chemglass CG-1949-X/CG-1969-M manifold systems, stainless steel NPT adapters and hoses with NPT threads. Temperature range -74° C to 318° C.
Catalog No. 50-131-1161 Supplier Chemglass Life Sciences Supplier No. CG1963T10
Thread Sealant, with PTFE, 2oz Tube. Smooth, nontoxic, paste forms a leak tight seal on pipe threads. Ideal for use with Chemglass CG-1949-X/CG-1969-M manifold systems, stainless steel NPT adapters and hoses with NPT threads. Temperature range -74° C to 318° C.