Learn More
COLDER PRODUCTS COMPANY Insert Drum CPC Polyethylene With 5cm (2 in ) American Buttress thread EPDM O-Rings (North American plastic drums) Temp Range 29 deg to 49 deg C (-20 deg to 120 deg F) Pressure Range 0 to 45psig (0 to 3 1 bar)
Catalog No. 503131272
Encompass_Preferred
Insert Drum CPC Polyethylene With 5cm (2 in ) American Buttress thread EPDM O-Rings (North American plastic drums) Temp Range 29 deg to 49 deg C (-20 deg to 120 deg F) Pressure Range 0 to 45psig (0 to 3 1 bar)
Catalog No. 50-313-1272 Supplier COLDER PRODUCTS COMPANY Supplier No. DQPRO202BUT
Insert Drum CPC Polyethylene With 5cm (2 in ) American Buttress thread EPDM O-Rings (North American plastic drums) Temp Range 29 deg to 49 deg C (-20 deg to 120 deg F) Pressure Range 0 to 45psig (0 to 3 1 bar)