Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Threaded Cap
GREENER_CHOICESDP

Catalog No. 31502298 Shop All Kemtech Products
Change view
Click to view available options
Thread Size (Metric):
GL10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
31-502-298 GL10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-298 Supplier Synthware™ Supplier No. RC1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thread Cap For GL10

  • Threaded cap
  • For use with GL10

Specifications

Color Red
Material Nylon
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) GL10
Liner Material Teflon
Neck Finish C1 mm
Graduated No
For Use With (Equipment) GL10
Closure Liner Teflon
Closure Material Plastic
Closure Size GL10
Autoclavable Not Autoclavable
Shape Cylindrical
Disposable Reusable
Product Type Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.