Learn More
Savillex Corporation 33 mm PFA Closure for 60ml vials
SDP
Catalog No. NC0705980
Encompass
Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C). Knurled finish on the closure. Leakproof seal.
Catalog No. NC0705980 Supplier Savillex Corporation Supplier No. 60003301
Service temperature range of PFA is ‑328°F to 500°F (‑200°C to 260°C). Knurled finish on the closure. Leakproof seal.