Learn More
Analytical Sales & Services Threaded Guide Studs for 96 Position Radiochemistry Reactor
Catalog No. 5000806657
Encompass
Guide Studs for 96 Position Radiochemistry Reactor\n\n• Partially threaded\n• Aligns block components. Thread studs into the corners of the Base Vial Rack, then slide the Transfer Plate and Top Cover onto the studs for perfect alignment.\n• 6-32 X 1-1/2” Long\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: 4
Catalog No. 50-008-06657 Supplier Analytical Sales & Services Supplier No. TS146
Guide Studs for 96 Position Radiochemistry Reactor\n\n• Partially threaded\n• Aligns block components. Thread studs into the corners of the Base Vial Rack, then slide the Transfer Plate and Top Cover onto the studs for perfect alignment.\n• 6-32 X 1-1/2” Long\n• Use with Radiochemistry Reactors (96950) or Conversion Kit (96940)\n\nQty/Pkg: 4