Learn More
Heidolph™ Threaded Joint KS 64 for Laborota 20, Hei-VAP Industrial Rotary Evaporator
SDP
Catalog No. 13900906 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-906 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-906 Supplier Heidolph™ Supplier No. 036210250
Add to Cart
Add to Cart
Threaded joint KS 64 connects vacuum fitting, condenser, elbow, and expansion vessel.
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator
Specifications
Description Coated
Type Clamp
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov