Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, 316L Stainless Steel, Straight, Sanitary Clamp to Threaded Adapter, 1.5"" Clamp x 3/4"" NPT(M)
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3070076
Encompass
These threaded adapter clamps by NPT(M) sanitary fittings are ideal for quick and easily installation of your sanitary set-up to an NPT male pipe thread. The 316L stainless steel couplings are suitable for the pharmaceutical industry. They offer great chemical compatibility for use with corrosive acids and bases. Clamps and gaskets required for a complete system are sold separately.
Catalog No. NC1330425