Learn More
Grainger Threaded Shank Drill 1/4 Cobalt
Catalog No. 502764411
Encompass_Preferred
The Grainger 1/16" Threaded Shank Drill Bit is a high-speed steel bit with a 1/16" diameter and 1/4" shank. It has a threaded shank that fits into a drill chuck and is designed for use in metal, wood, and plastic. The 135 degree split point tip starts drilling with less pressure. Overall length is 2-1/2". Made in the USA.
Catalog No. 50-276-4411 Supplier Grainger Supplier No. 16W823
The Grainger 1/16" Threaded Shank Drill Bit is a high-speed steel bit with a 1/16" diameter and 1/4" shank. It has a threaded shank that fits into a drill chuck and is designed for use in metal, wood, and plastic. The 135 degree split point tip starts drilling with less pressure. Overall length is 2-1/2". Made in the USA.