Learn More
Grainger Threaded Shank Drill #11 Cobalt
Catalog No. 502763768
Encompass_Preferred
The Grainger 11/16" Threaded Shank Drill Bit is made of high speed steel and has a black oxide finish for durability. It features a threaded shank that fits power drills and can drill through metal, wood, and plastic. The drill bit has a 118 degree point angle and is 11/16" in diameter. It can drill holes up to 1/2" deep. The overall length of the drill bit is 2-1/2". This threaded drill bit is ideal for metalworking applications that require drilling precise holes.
Catalog No. 50-276-3768 Supplier Grainger Supplier No. 16W774
The Grainger 11/16" Threaded Shank Drill Bit is made of high speed steel and has a black oxide finish for durability. It features a threaded shank that fits power drills and can drill through metal, wood, and plastic. The drill bit has a 118 degree point angle and is 11/16" in diameter. It can drill holes up to 1/2" deep. The overall length of the drill bit is 2-1/2". This threaded drill bit is ideal for metalworking applications that require drilling precise holes.