Learn More
Grainger Threaded Shank Drill #12 Cobalt
Catalog No. 502764410
Encompass_Preferred
The Grainger 12-16 Threaded Shank Drill Bit is a high-speed steel bit with a 1/2" hex shank and a cutting diameter of 12-16". It has a 118 degree point angle and is designed for use in metalworking applications. The bit features a threaded shank that allows for quick changes between tools. Made in the USA, this drill bit can bore holes in steel, iron, bronze, aluminum and other metals. It is ideal for use in drill presses, magnetic base drills, and milling machines. With proper use, this durable drill bit provides accurate cuts and smooth hole boring.
Catalog No. 50-276-4410 Supplier Grainger Supplier No. 16W795
The Grainger 12-16 Threaded Shank Drill Bit is a high-speed steel bit with a 1/2" hex shank and a cutting diameter of 12-16". It has a 118 degree point angle and is designed for use in metalworking applications. The bit features a threaded shank that allows for quick changes between tools. Made in the USA, this drill bit can bore holes in steel, iron, bronze, aluminum and other metals. It is ideal for use in drill presses, magnetic base drills, and milling machines. With proper use, this durable drill bit provides accurate cuts and smooth hole boring.