Learn More
Grainger Threaded Shank Drill #12 Cobalt
Catalog No. 502764232
Encompass_Preferred
The Grainger 12-16W818 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw machine length drill point. It has a 118 degree point angle and is uncoated with an overall length of 1-1/2". The bit diameter is 1/8" and it can drill through metal, wood, and plastic. The maximum rpm is 2750. This threaded shank drill is designed for use in hand drills or drill presses.
Catalog No. 50-276-4232 Supplier Grainger Supplier No. 16W818
The Grainger 12-16W818 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw machine length drill point. It has a 118 degree point angle and is uncoated with an overall length of 1-1/2". The bit diameter is 1/8" and it can drill through metal, wood, and plastic. The maximum rpm is 2750. This threaded shank drill is designed for use in hand drills or drill presses.