Learn More
Grainger Threaded Shank Drill #13 Cobalt
Catalog No. 502764376
Encompass_Preferred
The Grainger 13/16" Threaded Shank Drill Bit is made of high speed steel and has a black oxide finish. It features a 135 degree split point tip for faster penetration and reduced walking. The bit has a 1/2" hex shank to fit power drills. It is 13/16" in diameter and 9-1/2" long overall. The drill bit is designed for drilling in metal and has a threaded shank that allows for quick bit changes. This heavy duty drill bit is ideal for industrial applications that require frequent drilling in thick metal.
Catalog No. 50-276-4376 Supplier Grainger Supplier No. 16W793
The Grainger 13/16" Threaded Shank Drill Bit is made of high speed steel and has a black oxide finish. It features a 135 degree split point tip for faster penetration and reduced walking. The bit has a 1/2" hex shank to fit power drills. It is 13/16" in diameter and 9-1/2" long overall. The drill bit is designed for drilling in metal and has a threaded shank that allows for quick bit changes. This heavy duty drill bit is ideal for industrial applications that require frequent drilling in thick metal.