Learn More
Grainger Threaded Shank Drill #16 Cobalt
Catalog No. 502764730
Encompass_Preferred
The Grainger 16-16W815 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point. It has a length of 2-1/2" and is designed for use in metal, wood, and plastic. The bit features a 135 degree split point tip that starts drilling faster with less walking or wandering. It can be used in hand drills or drill presses and is ideal for drilling pilot holes. The high-speed steel construction provides strength and durability. This threaded shank drill bit allows for quick bit changes and reduced wobble during use. Made in the USA.
Catalog No. 50-276-4730 Supplier Grainger Supplier No. 16W815
The Grainger 16-16W815 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point. It has a length of 2-1/2" and is designed for use in metal, wood, and plastic. The bit features a 135 degree split point tip that starts drilling faster with less walking or wandering. It can be used in hand drills or drill presses and is ideal for drilling pilot holes. The high-speed steel construction provides strength and durability. This threaded shank drill bit allows for quick bit changes and reduced wobble during use. Made in the USA.