Learn More
Grainger Threaded Shank Drill #20 HSS
Catalog No. 502763767
Encompass_Preferred
The Grainger 20-piece Threaded Shank Drill Bit Set includes high-speed steel drill bits with black oxide finish for durability. The set contains sizes ranging from 1/16 inch to 1/2 inch by 64ths. The drill bits have a threaded shank that fits power drills. This 20-piece set provides a range of drill bit sizes useful for metalworking applications like drilling holes in metal, wood, and plastic. The drill bits can be used with hand drills, drill presses, and milling machines.
Catalog No. 50-276-3767 Supplier Grainger Supplier No. 16W702
The Grainger 20-piece Threaded Shank Drill Bit Set includes high-speed steel drill bits with black oxide finish for durability. The set contains sizes ranging from 1/16 inch to 1/2 inch by 64ths. The drill bits have a threaded shank that fits power drills. This 20-piece set provides a range of drill bit sizes useful for metalworking applications like drilling holes in metal, wood, and plastic. The drill bits can be used with hand drills, drill presses, and milling machines.