Learn More
Grainger Threaded Shank Drill 3/16 Cobalt
Catalog No. 502764329
Encompass_Preferred
The Grainger COBALT THREADED SHANK DRILL is a long, 3" cobalt drill bit with a threaded shank. It has a 135 degree split point tip for fast, easy drilling and reduced walking. The cobalt alloy provides high speed capability and wear resistance. This USA-made drill bit is ideal for drilling in stainless steel, high tensile steels, and other hard materials. It has a 3/8"-24 threaded shank to fit power drills. Overall, this is a durable, high performance cobalt drill bit for heavy duty drilling applications.
Catalog No. 50-276-4329 Supplier Grainger Supplier No. 16W817
The Grainger COBALT THREADED SHANK DRILL is a long, 3" cobalt drill bit with a threaded shank. It has a 135 degree split point tip for fast, easy drilling and reduced walking. The cobalt alloy provides high speed capability and wear resistance. This USA-made drill bit is ideal for drilling in stainless steel, high tensile steels, and other hard materials. It has a 3/8"-24 threaded shank to fit power drills. Overall, this is a durable, high performance cobalt drill bit for heavy duty drilling applications.