Learn More
Grainger Threaded Shank Drill 3/16 HSS
Catalog No. 502764849
Encompass_Preferred
The Grainger 3/16" Threaded Shank Drill Bit is a high-speed steel bit with a threaded shank for use in hand drills or drill presses. It has a 118 degree point angle and is designed for drilling metal, wood, and plastics. The overall length is 2-1/2" and the shank diameter is 3/16". It can drill holes up to 3/16" in diameter. The bit features a black oxide finish for reduced friction and improved wear resistance. Made in the USA.
Catalog No. 50-276-4849 Supplier Grainger Supplier No. 16W728
The Grainger 3/16" Threaded Shank Drill Bit is a high-speed steel bit with a threaded shank for use in hand drills or drill presses. It has a 118 degree point angle and is designed for drilling metal, wood, and plastics. The overall length is 2-1/2" and the shank diameter is 3/16". It can drill holes up to 3/16" in diameter. The bit features a black oxide finish for reduced friction and improved wear resistance. Made in the USA.