Learn More
Grainger Threaded Shank Drill 3/8 Cobalt
Catalog No. 502764230
Encompass_Preferred
The Grainger 3/16" Threaded Shank Drill Bit is a high-speed steel bit with a threaded shank for use in hand drills or drill presses. It has a 118 degree point angle and is designed for drilling into metal, wood, and plastics. The overall length is 2-1/2" and it has a 3/16" diameter. The shank diameter is 1/4". The bit features a black oxide finish for reduced friction and improved wear resistance. Made in the USA.
Catalog No. 50-276-4230 Supplier Grainger Supplier No. 16W803
The Grainger 3/16" Threaded Shank Drill Bit is a high-speed steel bit with a threaded shank for use in hand drills or drill presses. It has a 118 degree point angle and is designed for drilling into metal, wood, and plastics. The overall length is 2-1/2" and it has a 3/16" diameter. The shank diameter is 1/4". The bit features a black oxide finish for reduced friction and improved wear resistance. Made in the USA.