Learn More
Grainger Threaded Shank Drill 3/8 Cobalt
Catalog No. 502763678
Encompass_Preferred
The Grainger 3/16" Threaded Shank Drill Bit is made of high-speed steel and has a split point tip for fast, clean hole cutting. It has a 3/16" diameter and is 3-1/2" long overall. The shank is threaded 1/4"-20 for use in drill presses. The drill bit can cut through mild steel, aluminum, wood, plastic and other materials. It features a bright finish for corrosion resistance. This USA-made drill bit is ideal for metalworking applications that require frequent hole drilling.
Catalog No. 50-276-3678 Supplier Grainger Supplier No. 16W780
The Grainger 3/16" Threaded Shank Drill Bit is made of high-speed steel and has a split point tip for fast, clean hole cutting. It has a 3/16" diameter and is 3-1/2" long overall. The shank is threaded 1/4"-20 for use in drill presses. The drill bit can cut through mild steel, aluminum, wood, plastic and other materials. It features a bright finish for corrosion resistance. This USA-made drill bit is ideal for metalworking applications that require frequent hole drilling.