Learn More
Grainger Threaded Shank Drill #30 Cobalt
Catalog No. 502764827
Encompass_Preferred
The Grainger 30-16W808 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point tip. It has a length of 2-1/2" and is designed for use in metal, wood, and plastic. The bit features a bright finish and a 135 degree split point for faster penetration and cleaner holes. Made in the USA, this threaded shank drill bit is ideal for power drills and drill presses.
Catalog No. 50-276-4827 Supplier Grainger Supplier No. 16W808
The Grainger 30-16W808 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and a #7 screw point tip. It has a length of 2-1/2" and is designed for use in metal, wood, and plastic. The bit features a bright finish and a 135 degree split point for faster penetration and cleaner holes. Made in the USA, this threaded shank drill bit is ideal for power drills and drill presses.