Learn More
Grainger Threaded Shank Drill #30 Cobalt
Catalog No. 502763769
Encompass_Preferred
The Grainger 16W785 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and #7 screw machine length. It has a 135 degree split point tip for faster penetration and cleaner holes. The bit diameter is 40/64" (5/8") and the overall length is 2-1/2". It can be used for drilling metal, wood, and plastic. Made in the USA.
Catalog No. 50-276-3769 Supplier Grainger Supplier No. 16W785
The Grainger 16W785 Threaded Shank Drill Bit is a high-speed steel bit with a 1/4" hex shank and #7 screw machine length. It has a 135 degree split point tip for faster penetration and cleaner holes. The bit diameter is 40/64" (5/8") and the overall length is 2-1/2". It can be used for drilling metal, wood, and plastic. Made in the USA.